Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Respiratory Syncytial Virus A2 Fusion Protein Antibody (R338), PerCP, Novus Biologicals™
SDP

Catalog No. NB306345 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB306345 0.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB306345 Supplier Novus Biologicals Supplier No. NBP289761PCP
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Human Respiratory Syncytial Virus A2 Fusion Protein Monoclonal antibody specifically detects Human Respiratory Syncytial Virus A2 Fusion Protein in Virus samples. It is validated for Western Blot, ELISA, Neutralization

Specifications

Antigen Human Respiratory Syncytial Virus A2 Fusion Protein
Applications Western Blot, ELISA, Neutralization
Classification Monoclonal
Clone R338
Conjugate PerCP
Dilution Western Blot, Sandwich ELISA Capture, Block/Neutralize
Formulation PBS
Gene Alias HRSVgp08, RSV, RSV fusion protein
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human Respiratory Syncytial Virus (Uniprot#: P03420.1; Met1-Thr529)
Purification Method Protein A purified
Quantity 0.2 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.