Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Respiratory Syncytial Virus Nucleoprotein VPN41 Antibody (4-18), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NBP250535AG Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP250535AG 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP250535AG Supplier Novus Biologicals Supplier No. NBP250535AF488
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Human Respiratory Syncytial Virus Nucleoprotein VPN41 Monoclonal antibody specifically detects Human Respiratory Syncytial Virus Nucleoprotein VPN41 in Virus samples. It is validated for Western Blot, ELISA.

Specifications

Antigen Human Respiratory Syncytial Virus Nucleoprotein VPN41
Applications Western Blot, ELISA
Classification Monoclonal
Clone 43208
Conjugate Alexa Fluor 488
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Host Species Mouse
Immunogen Gradient-purified RSN-2 virus (subgroup B) treated with 0.1% SDS at 100C for 2 mins.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Virology Bacteria and Parasites
Primary or Secondary Primary
Test Specificity This antibody reacts with both human RSV subgroups A and B.
Target Species Virus
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.