Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Respiratory Syncytial Virus Phosphoprotein Antibody (3-5-18), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NB273802 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB273802 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB273802 Supplier Novus Biologicals Supplier No. NBP250244AF594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Human Respiratory Syncytial Virus Phosphoprotein Monoclonal antibody specifically detects Human Respiratory Syncytial Virus Phosphoprotein in Virus samples. It is validated for Western Blot, ELISA, Immunofluorescence

Specifications

Antigen Human Respiratory Syncytial Virus Phosphoprotein
Applications Western Blot, ELISA, Immunofluorescence
Classification Monoclonal
Clone 2018-03-05 00:00:00
Conjugate Alexa Fluor 594
Dilution Western Blot, ELISA, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Host Species Mouse
Immunogen The immunogen was gradient-purified RSN-2 virus (subgroup B) that was then treated with 0.1% SDS at 100C for 2 min. The procedure used to produce this antibody is described in Gimenez et al. (1984).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.