Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Respiratory Syncytial Virus (RSV) A2 Fusion Protein Rabbit anti-Virus, Clone: R338, Novus Biologicals™
SDP

Catalog No. NB020998 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
200 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB020998 200 μg
NB020999 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB020998 Supplier Novus Biologicals Supplier No. NBP289761200UG
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Human Respiratory Syncytial Virus (RSV) A2 Fusion Protein Monoclonal specifically detects Human Respiratory Syncytial Virus (RSV) A2 Fusion Protein in Virus samples. It is validated for Sandwich ELISA Capture, Block/Neutralize.

Specifications

Antigen Human Respiratory Syncytial Virus (RSV) A2 Fusion Protein
Applications ELISA
Classification Monoclonal
Clone R338
Conjugate Unconjugated
Dilution Sandwich ELISA Capture 0.5-4 ug/ml
Formulation 0.2 um filtered solution in PBS
Gene Alias HRSVgp08, RSV, RSV fusion protein
Host Species Rabbit
Immunogen Recombinant Human Respiratory Syncytial Virus (RSV) A2 Fusion Protein protein
Purification Method Protein A purified
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.