Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human RIPK3/RIP3 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
$200.16 - $472.87
Specifications
| Antigen | RIPK3/RIP3 |
|---|---|
| Clone | 1312D |
| Dilution | Western Blot 2 μg/mL, Immunocytochemistry 1-25 μg/mL |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB76042SP
![]() |
R&D Systems
MAB76042SP |
25 μg |
Each for $200.16
|
|
|||||
MAB76042100
![]() |
R&D Systems
MAB76042100 |
100 μg |
Each for $472.87
|
|
|||||
Description
RIPK3/RIP3 Monoclonal specifically detects RIPK3/RIP3 in Human samples. It is validated for Western Blot, Immunocytochemistry.Specifications
| RIPK3/RIP3 | |
| Western Blot 2 μg/mL, Immunocytochemistry 1-25 μg/mL | |
| Monoclonal | |
| Lyophilized | |
| Human | |
| Q9Y572 | |
| 11035 | |
| IgG | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
| 1312D | |
| Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| EC 2.7.11.1, Receptor Interacting Serine/Threonine Kinase 3, Receptor-interacting protein 3, receptor-interacting serine/threonine-protein kinase 3, receptor-interacting serine-threonine kinase 3, RIP3, RIP-3, RIP-like protein kinase 3 | |
| Synthetic peptide containing human RIPK3/RIP3, Accession # Q9Y572 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human RIPK3/RIP3 in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title