Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human SCL/Tal1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $468.00
Specifications
Antigen | SCL/Tal1 |
---|---|
Clone | 333803 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL, Immunocytochemistry 8-25 ug/mL |
Applications | Western Blot, Immunocytochemistry |
Description
SCL/Tal1 Monoclonal specifically detects SCL/Tal1 in Human samples. It is validated for Western Blot, Immunocytochemistry.Specifications
SCL/Tal1 | |
LYOPH | |
Western Blot, Immunocytochemistry | |
Unconjugated | |
Mouse | |
Human | |
P17542 | |
6886 | |
E. coli-derived recombinant human SCL/Tal1 Arg199-Gly266 Accession # P17542 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human SCL/Tal1 in direct ELISAs and Western blots. |
333803 | |
Western Blot 1 ug/mL, Immunocytochemistry 8-25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
BHLHA17, bHLHa17tal-1, Class A basic helix-loop-helix protein 17, SCLT-cell acute lymphocytic leukemia protein 1, Stem cell protein, Tal1, TAL-1, T-cell acute lymphocytic leukemia 1, T-cell leukemia/lymphoma protein 5, TCL5, TCL5tal-1 product | |
TAL1 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title