Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Serum Albumin Antibody, Novus Biologicals™
SDP

Catalog No. NBP221669 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP221669 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP221669 Supplier Novus Biologicals Supplier No. NBP221669
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Human Serum Albumin Polyclonal specifically detects Human Serum Albumin in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Human Serum Albumin
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:2000-1:10000, ELISA 1:20000-1:100000, Immunohistochemistry 1:1000-1:5000, Immunohistochemistry-Paraffin 1:1000-1:5000
Formulation Lyophilized from Antiserum with 0.01% Sodium Azide
Gene Accession No. P02768
Gene Alias albumin, cell growth inhibiting protein 42, DKFZp779N1935, growth-inhibiting protein 20, PRO0883, PRO0903, PRO1341, serum albumin
Gene Symbols ALB
Host Species Rabbit
Immunogen Albumin (Human Serum) (Uniprot: P02768)
Molecular Weight of Antigen 69.37 kDa
Purification Method Delipidation and Defibrination
Quantity 2 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Golgi Apparatus Markers, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 213
Test Specificity Cross reactivity against Albumin from other tissues and species may occur but has not been specifically determined.
Reconstitution Reconstitute with 2.0 ml deionized water (or equivalent)
Target Species Human
Content And Storage Store lyophilized antibody at 4°C. Aliquot reconstituted liquid and store at -20°C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.