Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Siglec-9 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 3 publications
$222.00 - $2070.00
Specifications
Antigen | Siglec-9 |
---|---|
Clone | 191240 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, Neutralization 0.015-0.075 ug/mL, CyTOF-ready, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
Applications | Western Blot, Flow Cytometry, Neutralization, CyTOF, ELISA |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB1139SP
![]() |
R&D Systems
MAB1139SP |
25 μg |
Each for $222.00
|
|
|||||
MAB1139100
![]() |
R&D Systems
MAB1139100 |
100 μg |
Each for $582.50
|
|
|||||
MAB1139500
![]() |
R&D Systems
MAB1139500 |
500 μg |
Each for $2,070.00
|
|
|||||
Description
Siglec-9 Monoclonal specifically detects Siglec-9 in Human samples. It is validated for Western Blot, Flow Cytometry, Neutralization, CyTOF-ready, ELISA Capture (Matched Antibody Pair).Specifications
Siglec-9 | |
LYOPH | |
Western Blot, Flow Cytometry, Neutralization, CyTOF, ELISA | |
Unconjugated | |
Mouse | |
Human | |
Q9Y336 | |
27180 | |
Mouse myeloma cell line NS0-derived recombinant human Siglec-9 Gln18-Gly348 (predicted) Accession # Q9Y336 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human Siglec-9 in direct ELISAs and Western blots. In direct ELISAs, less than 10% cross-reactivity with recombinant mouse Siglec-E is observed and less than 1% cross-reactivity with recombinant human (rh) Siglec-3, rhSiglec-5, rhSiglec-6, rhSiglec-7, rhSiglec-8 or rhSiglec-10 is observed. In Western blots, approximately 100% cross-reactivity with recombinant mouse Siglec-E is observed under non-reduced conditions. |
191240 | |
Western Blot 1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, Neutralization 0.015-0.075 ug/mL, CyTOF-ready, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CD329, CDw329, FOAP-9, OBBP-Like, Protein FOAP-9, sialic acid binding Ig-like lectin 9, sialic acid-binding Ig-like lectin 9, siglec-9, Siglec9 | |
SIGLEC9 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title