Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Slug Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB7408SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB7408SP 25 μg
MAB7408100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB7408SP Supplier R&D Systems Supplier No. MAB7408SP
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Slug Monoclonal specifically detects Slug in Human samples. It is validated for Intracellular Staining by Flow Cytometry, CyTOF-ready.

Specifications

Antigen Slug
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 666633
Conjugate Unconjugated
Dilution Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready
Formulation Lyophilized from a 0.2 µm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS.
Gene Accession No. O43623
Gene Alias MGC10182, Neural crest transcription factor Slug, Protein snail homolog 2, slug homolog, zinc finger protein (chicken), SLUGH, SLUGH1, SLUGzinc finger protein, SNAI2, snail 2, snail homolog 2 (Drosophila), SNAIL2, WS2D, zinc finger protein SNAI2
Host Species Mouse
Immunogen E. coli-derived recombinant human Slug, Met1-His268, Accession # O43623
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6591
Test Specificity Detects human Slug in direct ELISAs.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.