Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human SOD1/Cu-Zn SOD Protein
SDP

Catalog No. p-200003770 Shop All R&D Systems Products
Click to view available options
Quantity:
0.02 mg
0.1mg
0.5 mg

Highly purified and high bioactivity. Generating reliable and reproducible results.

Specific activity is > 500 units/mg, in which one unit will inhibit the rate of reduction of cytochrome c by 50% in a coupled system, using xanthine and Xanthine oxidase at pH 7.5 at 25C.

Specifications

Accession Number P22309
For Use With (Application) Bioactivity, SDS-PAGE
Formulation 20mM Tris buffer (pH 7.5), 10% glycerol
Gene ID (Entrez) 6647
Molecular Weight (g/mol) 16kDa
Name SOD1/Cu-Zn SOD Protein
Purification Method Protein
Quantity 0.1mg
Source E. Coli
Immunogen SOD1, 1-154aa, Human, Recombinant, E.coli (NP_000454).
Storage Requirements −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Purity or Quality Grade >95%, by SDS-PAGE
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.