Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human SREC-II/SCARF2 Antibody, R&D Systems™

Goat Polyclonal Antibody
$200.50 - $468.50
Specifications
Antigen | SREC-II/SCARF2 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL, Blockade of Receptor-ligand Interaction |
Applications | Western Blot, Blockade of Receptor-ligand Interaction |
Classification | Polyclonal |
Description
SREC-II/SCARF2 Polyclonal specifically detects SREC-II/SCARF2 in Human samples. It is validated for Western Blot, Blockade of Receptor-ligand Interaction.Specifications
SREC-II/SCARF2 | |
Western Blot 0.1 ug/mL, Blockade of Receptor-ligand Interaction | |
Polyclonal | |
Goat | |
Human | |
HUMZD58C02, SCARF2, scavenger receptor class F, member 2, Scavenger receptor expressed by endothelial cells 2 protein, SREC2NSR1, SRECII, SREC-IIscavenger receptor class F member 2, SRECRP-1, SREPCR | |
SCARF2 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot, Blockade of Receptor-ligand Interaction | |
Unconjugated | |
RUO | |
Q96GP6 | |
91179 | |
Mouse myeloma cell line NS0-derived recombinant human SREC-II/SCARF2 Ala42-Gly442 Accession # Q96GP6 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects SREC-II/SCARF2 in Western blots. In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant human SREC-I is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title