Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human ST2/IL-33R PE-conjugated Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 4 publications
Specifications
Antigen | ST2/IL-33R |
---|---|
Dilution | Flow Cytometry 10 uL/10^6 cells |
Applications | Flow Cytometry |
Classification | Polyclonal |
Conjugate | R-PE |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
FAB5231P025
|
R&D Systems
FAB5231P025 |
25 Tests | N/A | N/A | N/A | ||||
FAB5231P100
|
R&D Systems
FAB5231P100 |
100 Tests | N/A | N/A | N/A | ||||
Description
ST2/IL-33R Polyclonal specifically detects ST2/IL-33R in Human samples. It is validated for Flow Cytometry.Specifications
ST2/IL-33R | |
Flow Cytometry | |
R-PE | |
RUO | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
DER4, DER4ST2growth stimulation-expressed, FIT-1, IL-1 R4, IL1R4, IL-1R4, IL-1RL1, IL-33 R, IL33R, IL-33R, interleukin 1 receptor-like 1, interleukin 1 receptor-related protein, interleukin-1 receptor-like 1, Ly84, MGC32623, Protein ST2, ST2L, ST2V, T1, T1homolog of mouse growth stimulation-expressed | |
IL1RL1 | |
IgG | |
Affinity Purified | |
Detects human ST2/IL-33 R in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human (rh) IL-1 RI, rhIL-1 RII, rhIL-18 RI, and rhST-7 is observed. |
Flow Cytometry 10 uL/10^6 cells | |
Polyclonal | |
Goat | |
Human | |
BAA02233 | |
9173 | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human ST2/IL-33 R Lys19-Phe328 Accession # BAA02233 | |
Primary | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title