Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Stanniocalcin 1/STC-1 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 3 publications
Supplier: R&D Systems AF2958
This item is not returnable.
View return policy
Description
Stanniocalcin 1/STC-1 Polyclonal specifically detects Stanniocalcin 1/STC-1 in Human samples. It is validated for Western Blot.Specifications
Stanniocalcin 1/STC-1 | |
Polyclonal | |
Unconjugated | |
P52823 | |
STC1 | |
Mouse myeloma cell line NS0-derived recombinant human Stanniocalcin 1 Thr18-Ala247 Accession # P52823 | |
100 μg | |
Primary | |
Detects human Stanniocalcin 1/STC-1 in direct ELISAs and Western blots. In these formats, less than 1% cross-reactivity with recombinant human STC-2 is observed. | |
Human | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 0.1 ug/mL | |
stanniocalcin 1, stanniocalcin-1, STC1, STC-1, STCSTC-1 | |
Goat | |
Affinity Purified | |
RUO | |
6781 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction