Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Stanniocalcin 1/STC-1 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 3 publications
$208.00 - $570.50
Specifications
Antigen | Stanniocalcin 1/STC-1 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Description
Stanniocalcin 1/STC-1 Polyclonal specifically detects Stanniocalcin 1/STC-1 in Human samples. It is validated for Western Blot.Specifications
Stanniocalcin 1/STC-1 | |
Western Blot 0.1 ug/mL | |
Polyclonal | |
Goat | |
Human | |
stanniocalcin 1, stanniocalcin-1, STC1, STC-1, STCSTC-1 | |
STC1 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot | |
Unconjugated | |
RUO | |
P52823 | |
6781 | |
Mouse myeloma cell line NS0-derived recombinant human Stanniocalcin 1 Thr18-Ala247 Accession # P52823 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human Stanniocalcin 1/STC-1 in direct ELISAs and Western blots. In these formats, less than 1% cross-reactivity with recombinant human STC-2 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title