Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Syndecan-3 Isoform 1 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
$200.50 - $549.50
Specifications
Antigen | SDC3 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Immunocytochemistry 5-15 ug/mL, CyTOF-ready |
Applications | Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF |
Classification | Polyclonal |
Description
SDC3 Polyclonal specifically detects SDC3 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF-ready.Specifications
SDC3 | |
Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Immunocytochemistry 5-15 ug/mL, CyTOF-ready | |
Polyclonal | |
Goat | |
Human | |
KIAA0468, N-syndecan, SDC3, SDCN, SYND3syndecan proteoglycan 3, syndecan 3, syndecan 3 (N-syndecan), syndecan neural type, syndecan-3, Syndecan3 | |
SDC3 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF | |
Unconjugated | |
RUO | |
O75056 | |
9672 | |
Mouse myeloma cell line NS0-derived recombinant human Syndecan-3 Gln48-Lys383 Accession # O75056 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human Syndecan-3 in direct ELISAs and Western blots. In direct ELISAs, approximately 40% cross-reactivity with recombinant mouse Syndecan-3 is observed and less than 1% cross-reactivity with recombinant mouse Syndecan-4 and recombinant human Syndecan-2 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title