Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human TAFA1/FAM19A1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB5154SP
This item is not returnable.
View return policy
Description
TAFA1/FAM19A1 Monoclonal specifically detects TAFA1/FAM19A1 in Human samples. It is validated for Western Blot.Specifications
| TAFA1/FAM19A1 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| Q7Z5A9 | |
| FAM19A1 | |
| E. coli-derived recombinant human TAFA1/FAM19A1 Ser26-Thr133 Accession # Q7Z5A9 | |
| 25 μg | |
| Primary | |
| Detects human TAFA1/FAM19A1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human TAFA2, 3, 4, or 5 is observed. | |
| Human | |
| Purified |
| Western Blot | |
| 492821 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Chemokine-like protein TAFA-1, FAM19A1, family with sequence similarity 19 (chemokine (C-C motif)-like), member A1, TAFA1, TAFA-1 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 407738 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction