Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human TAFA1/FAM19A1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$188.28 - $531.97
Specifications
| Antigen | TAFA1/FAM19A1 |
|---|---|
| Clone | 492821 |
| Concentration | LYOPH |
| Dilution | Western Blot 1 ug/mL |
| Applications | Western Blot |
Description
TAFA1/FAM19A1 Monoclonal specifically detects TAFA1/FAM19A1 in Human samples. It is validated for Western Blot.Specifications
| TAFA1/FAM19A1 | |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q7Z5A9 | |
| 407738 | |
| E. coli-derived recombinant human TAFA1/FAM19A1 Ser26-Thr133 Accession # Q7Z5A9 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human TAFA1/FAM19A1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human TAFA2, 3, 4, or 5 is observed. |
| 492821 | |
| Western Blot 1 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Chemokine-like protein TAFA-1, FAM19A1, family with sequence similarity 19 (chemokine (C-C motif)-like), member A1, TAFA1, TAFA-1 | |
| FAM19A1 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title