Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Thymidine Kinase 1 Antibody, R&D Systems™

Sheep Antibody
$200.50 - $549.50
Specifications
Antigen | Thymidine Kinase 1 |
---|---|
Dilution | Simple Western 20 ug/mL, Immunohistochemistry 1-25 ug/mL |
Applications | Western Blot, Immunohistochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
Thymidine Kinase 1 specifically detects Thymidine Kinase 1 in Human samples. It is validated for Simple Western, Immunohistochemistry.Specifications
Thymidine Kinase 1 | |
Western Blot, Immunohistochemistry | |
Unconjugated | |
Sheep | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
EC 2.7.1.21, thymidine kinase 1 soluble isoform, thymidine kinase 1, soluble, thymidine kinase, cytosolic, thymidine kinase-1, TK1, TK2 | |
E. coli-derived human Thymidine Kinase 1 protein, Ser2-Asn234, Accession No. P04183 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human Thymidine Kinase 1 Protein in direct ELISAs. |
Simple Western 20 ug/mL, Immunohistochemistry 1-25 ug/mL | |
Polyclonal | |
Lyophilized | |
Human | |
P04183 | |
7083.0 | |
IgG | |
Antigen Affinity-purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title