Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human Total MDM2/HDM2 DuoSet IC ELISA

Catalog No. p-4754559
Click to view available options
Packaging Quantity:
15 Plates
2 Plates
5 Plates
This item is not returnable. View return policy
This item is not returnable. View return policy

Contains the basic components required for the development of sandwich ELISAs to measure in cell lysates.

The Human Total MDM2/HDM2 DuoSet IC ELISA is a Solid Phase Sandwich ELISA that quantifies human MDM2/HDM2.

Specifications

Product Type ELISA Development Kit for Intracellular Proteins
Quantity 1 Kit (for 2 Plates)
Species Human
Conjugate Biotin
Assay Range 312.00 to 20000 pg/mL
Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules
For Use With (Application) ELISA
Gene ID (Entrez) 4193
Synonym ACTFS, HDM2, HDMX, MDM2 oncogene, E3 ubiquitin protein ligase
Packaging Quantity 2 Plates
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.