Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human Total VEGF R3/Flt-4 DuoSet IC

Catalog No. p-4754592
Click to view available options
Packaging Quantity:
15 Plates
2 Plates
5 Plates
This item is not returnable. View return policy
This item is not returnable. View return policy

Contains the basic components required for the development of sandwich ELISAs to measure in cell lysates.

  • Optimized capture and detection antibody pairings with recommended concentrations save lengthy development time
  • Development protocols are provided to guide further assay optimization
  • Assay can be customized to your specific needs

Specifications

Product Type ELISA Development Kit for Intracellular Proteins
Quantity 1 Kit
Species Human
Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules
Gene ID (Entrez) 2324
Packaging Quantity 15 Plates
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.