Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human TRA-1-81 PE-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. p-200041088
Change view
Click to view available options
Quantity:
25 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB8495P025 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB8495P025 Supplier R&D Systems Supplier No. FAB8495P025
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 2 publications

TRA-1-81 Monoclonal specifically detects TRA-1-81 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen TRA-1-81
Applications Flow Cytometry
Classification Monoclonal
Clone TRA-1-81
Conjugate R-PE
Dilution Flow Cytometry 10 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Alias gp200, MGC138240, PC, PCLP, PODXL, TRA181
Gene Symbols PODXL
Host Species Mouse
Immunogen Human embryonal carcinoma cell line 2102Ep
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Detects human TRA-1-81 in flow cytometry.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.