Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Transferrin Antibody, R&D Systems™

Goat Antibody
$188.28 - $579.81
Specifications
| Antigen | Transferrin |
|---|---|
| Dilution | Western Blot 0.1 ug/mL |
| Applications | Western Blot |
| Conjugate | Unconjugated |
| Host Species | Goat |
Description
Transferrin specifically detects Transferrin in Human samples. It is validated for Western Blot.Specifications
| Transferrin | |
| Western Blot | |
| Goat | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| 7018 | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot 0.1 ug/mL | |
| Unconjugated | |
| Human | |
| ApoTransferrin, Beta-1 metal-binding globulin, DKFZp781D0156, EC 3.4.21, HoloTransferrin, PRO1557, PRO2086, serotransferrin, Siderophilin, TF, transferrin | |
| Human plasma-derived | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects Human Transferrin in ELISAs. Detects human Transferrin and Apo-Transferrin in Western blots. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title