Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human TREM-1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. p-7235417
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB12781SP 25 μg
MAB12781100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB12781SP Supplier R&D Systems Supplier No. MAB12781SP
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

TREM1 Monoclonal specifically detects TREM1 in Human samples. It is validated for Flow Cytometry, ELISA, CyTOF-ready.

Specifications

Antigen TREM1
Applications Flow Cytometry, ELISA, CyTOF
Classification Monoclonal
Clone 888111
Conjugate Unconjugated
Dilution Flow Cytometry 0.25 ug/10^6 cells, ELISA, CyTOF-ready
Formulation Lyophilized from a 0.2 µm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS.
Gene Accession No. Q9NP99
Gene Alias CD354, CD354 antigen, TREM1, TREM-1Triggering receptor expressed on monocytes 1, triggering receptor expressed on myeloid cells 1, triggering-receptor TREM1
Host Species Mouse
Immunogen E. coli-derived recombinant human TREM-1, Ala21-Asn150, Accession # Q9NP99
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54210
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.