Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human TSG101 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$184.70 - $454.00
Specifications
Antigen | TSG101 |
---|---|
Clone | 1065916 |
Dilution | Western Blot 2 μg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB114381SP
![]() |
R&D Systems
MAB114381SP |
25 μg |
Each for $184.70
|
|
|||||
RD10000180
![]() |
R&D Systems
MAB114381100 |
100 μg |
Each for $454.00
|
|
|||||
Description
TSG101 Monoclonal antibody specifically detects TSG101 in Human samples. It is validated for Western BlotSpecifications
TSG101 | |
Western Blot 2 μg/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
ESCRT-I complex subunit TSG101, TSG10, tumor susceptibility gene 10, tumor susceptibility gene 101, tumor susceptibility gene 101 protein, tumor susceptibility protein, VPS23 | |
TSG101 containing synthetic peptide, Accession # Q99816 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. |
1065916 | |
Western Blot | |
Unconjugated | |
Mouse | |
Human | |
Q99816 | |
7251 | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title