Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human TSHR Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB65341SP
This item is not returnable.
View return policy
Description
TSHR Monoclonal specifically detects TSHR in Human samples. It is validated for ELISA.Specifications
TSHR | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in TBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
CHNG1, LGR3, LGR3hTSHR-I, MGC75129, seven transmembrane helix receptor, thyroid stimulating hormone receptor, Thyroid-stimulating hormone receptor, thyrotropin receptor, thyrotropin receptor-I, hTSHR-I, TSH R, TSH-R | |
NS0 mouse myeloma cell line transfected with human TSH R Accession # P16473 | |
25 μg | |
Primary | |
Detects human TSH R in direct ELISAs. | |
Human | |
Purified |
ELISA | |
484416 | |
ELISA | |
P16473 | |
Mouse | |
Protein A or G purified from ascites | |
RUO | |
7253 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG3 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
RUO
Spot an opportunity for improvement?Share a Content Correction