Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human TSHR Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $526.00
Specifications
Antigen | TSHR |
---|---|
Clone | 484416 |
Dilution | ELISA |
Applications | ELISA |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB65341SP
![]() |
R&D Systems
MAB65341SP |
25 μg |
Each for $200.50
|
|
|||||
MAB65341100
![]() |
R&D Systems
MAB65341100 |
100 μg |
Each for $526.00
|
|
|||||
Description
TSHR Monoclonal specifically detects TSHR in Human samples. It is validated for ELISA.Specifications
TSHR | |
ELISA | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in TBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
CHNG1, LGR3, LGR3hTSHR-I, MGC75129, seven transmembrane helix receptor, thyroid stimulating hormone receptor, Thyroid-stimulating hormone receptor, thyrotropin receptor, thyrotropin receptor-I, hTSHR-I, TSH R, TSH-R | |
NS0 mouse myeloma cell line transfected with human TSH R Accession # P16473 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human TSH R in direct ELISAs. |
484416 | |
ELISA | |
Unconjugated | |
Mouse | |
Human | |
P16473 | |
7253 | |
IgG3 | |
Protein A or G purified from ascites | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
RUO
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title