Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human UNC13B Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB6188SP
This item is not returnable.
View return policy
Description
UNC13B Monoclonal specifically detects UNC13B in Human samples. It is validated for Immunohistochemistry.Specifications
UNC13B | |
Monoclonal | |
LYOPH | |
Immunohistochemistry 8-25 ug/mL | |
O14795 | |
UNC13B | |
E. Coli-derived recombinant human UNC13B Asp418-Ser1591 Accession # O14795 | |
25 μg | |
Primary | |
Detects human UNC13B in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) UNC13A and rhUNC13C is observed. | |
Human | |
Purified |
Immunohistochemistry | |
584102 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
hmunc13, homolog of rat Munc13 (diacylglycerol-binding), MGC133279, MGC133280, MUNC13, Munc13-2, MUNC13B, protein unc-13 homolog B, unc-13 homolog B (C. elegans), Unc13h2, unc-13-like (C. elegans), UNC13Unc13h2 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
10497 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction