Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human UNC13B Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $502.50
Specifications
Antigen | UNC13B |
---|---|
Clone | 584102 |
Concentration | LYOPH |
Dilution | Immunohistochemistry 8-25 ug/mL |
Applications | Immunohistochemistry |
Description
UNC13B Monoclonal specifically detects UNC13B in Human samples. It is validated for Immunohistochemistry.Specifications
UNC13B | |
LYOPH | |
Immunohistochemistry | |
Unconjugated | |
Mouse | |
Human | |
O14795 | |
10497 | |
E. Coli-derived recombinant human UNC13B Asp418-Ser1591 Accession # O14795 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human UNC13B in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) UNC13A and rhUNC13C is observed. |
584102 | |
Immunohistochemistry 8-25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
hmunc13, homolog of rat Munc13 (diacylglycerol-binding), MGC133279, MGC133280, MUNC13, Munc13-2, MUNC13B, protein unc-13 homolog B, unc-13 homolog B (C. elegans), Unc13h2, unc-13-like (C. elegans), UNC13Unc13h2 | |
UNC13B | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title