Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human UNC5H3 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $655.00
Specifications
Antigen | UNC5H3/UNC5C |
---|---|
Clone | 200414 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL, Immunohistochemistry 5-25 ug/mL |
Applications | Western Blot, Immunohistochemistry |
Description
UNC5H3/UNC5C Monoclonal specifically detects UNC5H3/UNC5C in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
UNC5H3/UNC5C | |
LYOPH | |
Western Blot, Immunohistochemistry | |
Unconjugated | |
Mouse | |
Human | |
O95185 | |
8633 | |
Mouse myeloma cell line NS0-derived recombinant human UNC5H3 Ala40-Asp376 Accession # O95185 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human UNC5H3 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human UNC5H4 or recombinant rat UNC5H2 is observed. |
200414 | |
Western Blot 1 ug/mL, Immunohistochemistry 5-25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
netrin receptor UNC5C, Protein unc-5 homolog 3, Protein unc-5 homolog C, unc-5 homolog 3, unc-5 homolog C (C. elegans), UNC5C, UNC5H3unc5 (C.elegans homolog) c | |
UNC5C | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title