Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human UNC5H4 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 3 publications
$200.50 - $549.50
Specifications
Antigen | UNC5H4/UNC5D |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Description
UNC5H4/UNC5D Polyclonal specifically detects UNC5H4/UNC5D in Human samples. It is validated for Western Blot.Specifications
UNC5H4/UNC5D | |
Western Blot 0.1 ug/mL | |
Polyclonal | |
Goat | |
Human | |
FLJ16019, KIAA1777Unc5h4, netrin receptor UNC5D, netrin receptor Unc5h4, PRO34692, Protein unc-5 homolog 4, Protein unc-5 homolog D, unc-5 homolog 4, unc-5 homolog D (C. elegans), UNC5D, UNC5H4 | |
UNC5D | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot | |
Unconjugated | |
RUO | |
NP_543148 | |
137970 | |
Mouse myeloma cell line NS0-derived recombinant human UNC5H4 Gly15-Ser373 Accession # NP_543148 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human UNC5H4 in direct ELISAs and Western blots. In these formats, less than 5% cross-reactivity with recombinant rat (rr) UNC5H1, rrUNC5H2, and recombinant human UNC5H3 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title