Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human VAP-1/AOC3 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
Supplier: R&D Systems MAB3957SP
This item is not returnable.
View return policy
Description
VAP-1/AOC3 Monoclonal specifically detects VAP-1/AOC3 in Human samples. It is validated for Western Blot.Specifications
VAP-1/AOC3 | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL | |
Q16853 | |
AOC3 | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human VAP-1/AOC3 Gly27-Asn763 Accession # Q16853 | |
25 μg | |
Primary | |
Detects human VAP-1/AOC3 in direct ELISAs and Western blots. In Western blots, 50% cross-reactivity with recombinant mouse VAP-1/AOC3 is observed. | |
Human | |
Purified |
Western Blot | |
393112 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
amine oxidase, copper containing 3 (vascular adhesion protein 1), AOC3, Copper amine oxidase, EC 1.4.3, HPAO, HPAOSSAO, Semicarbazide-sensitive amine oxidase, SSAO, VAP1, VAP1EC 1.4.3.21, VAP-1membrane primary amine oxidase, Vascular adhesion protein 1 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
8639 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction