Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human VAP-1/AOC3 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
$200.50 - $514.50
Specifications
Antigen | VAP-1/AOC3 |
---|---|
Clone | 393112 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL |
Applications | Western Blot |
Description
VAP-1/AOC3 Monoclonal specifically detects VAP-1/AOC3 in Human samples. It is validated for Western Blot.Specifications
VAP-1/AOC3 | |
LYOPH | |
Western Blot | |
Unconjugated | |
Mouse | |
Human | |
Q16853 | |
8639 | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human VAP-1/AOC3 Gly27-Asn763 Accession # Q16853 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human VAP-1/AOC3 in direct ELISAs and Western blots. In Western blots, 50% cross-reactivity with recombinant mouse VAP-1/AOC3 is observed. |
393112 | |
Western Blot 1 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
amine oxidase, copper containing 3 (vascular adhesion protein 1), AOC3, Copper amine oxidase, EC 1.4.3, HPAO, HPAOSSAO, Semicarbazide-sensitive amine oxidase, SSAO, VAP1, VAP1EC 1.4.3.21, VAP-1membrane primary amine oxidase, Vascular adhesion protein 1 | |
AOC3 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title