Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human VE-Cadherin APC-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. p-4712157
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB9381A025 25 Tests
FAB9381A100 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. FAB9381A025 Supplier R&D Systems Supplier No. FAB9381A025
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 1 publication

VE-Cadherin Monoclonal specifically detects VE-Cadherin in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen VE-Cadherin
Applications Flow Cytometry
Classification Monoclonal
Clone 123413
Conjugate APC
Dilution Flow Cytometry 10 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. P33151
Gene Alias 7B4, 7B4 antigen, cadherin 5, type 2 (vascular endothelium), cadherin 5, type 2, VE-cadherin (vascular epithelium), cadherin-5, CD144, CD144 antigen, CDH5, endothelial-specific cadherin, FLJ17376, Vascular endothelial cadherin, VE-cadherin, VECadherin
Gene Symbols CDH5
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human VE-Cadherin Asp48-Gln593 Accession # P33151
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1003
Test Specificity Detects human VE-Cadherin in Western blots. In Western blots, 25% cross-reactivity with recombinant mouse VE-Cadherin and no cross-reactivity with recombinant human (rh) Cadherin-17 or rhP-Cadherin is observed.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.