Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human VSIG2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $467.50
Specifications
Antigen | VSIG2 |
---|---|
Clone | 541522 |
Dilution | Flow Cytometry 0.25 μg/106 cells, CyTOF-ready |
Applications | Flow Cytometry, CyTOF |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB51771SP
![]() |
R&D Systems
MAB51771SP |
25 μg |
Each for $200.50
|
|
|||||
MAB51771100
![]() |
R&D Systems
MAB51771100 |
100 μg |
Each for $467.50
|
|
|||||
Description
VSIG2 Monoclonal specifically detects VSIG2 in Human samples. It is validated for Flow Cytometry, CyTOF-ready.Specifications
VSIG2 | |
Flow Cytometry 0.25 μg/106 cells, CyTOF-ready | |
Monoclonal | |
Mouse | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
2210413P10Rik, Cortical thymocyte-like protein, CTH, CTHCTXLcortical thymocyte receptor (X. laevis CTX) like, CT-like protein, CTXL, V-set and immunoglobulin domain containing 2, V-set and immunoglobulin domain-containing protein 2 | |
Mouse myeloma cell line NS0-derived recombinant human VSIG2, Val24-Ala243, Accession # Q961Q7-1 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human VSIG2 in direct ELISAs. |
541522 | |
Flow Cytometry, CyTOF | |
Unconjugated | |
Human | |
Q961Q7-1 | |
23584 | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title