Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Humanin Antibody, Biotin, Novus Biologicals™
SDP

Catalog No. NB210741 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB210741 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB210741 Supplier Novus Biologicals Supplier No. NB300245B
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Humanin Polyclonal antibody specifically detects Humanin in Human samples. It is validated for Western Blot

Specifications

Antigen Humanin
Applications Western Blot
Classification Polyclonal
Conjugate Biotin
Formulation PBS
Gene Alias HN, HUNIN
Host Species Rabbit
Immunogen Residues 9-24 (LLLLTSEIDLPVKRRA) of the human Humanin protein.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100462977
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.