Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Humanin Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB300245 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB300245 0.1 mL
NB018152 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB300245 Supplier Novus Biologicals Supplier No. NB300245
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Humanin Polyclonal specifically detects Humanin in Human samples. It is validated for Western Blot.

Specifications

Antigen Humanin
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500
Formulation PBS with 0.02% Sodium Azide
Gene Alias HN, HUNIN
Gene Symbols MTRNR2L1
Host Species Rabbit
Immunogen Residues 9-24 (LLLLTSEIDLPVKRRA) of the human Humanin protein.
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100462977
Test Specificity This is specific for Humanin.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.