Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Humanin Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.50 - $499.50
Specifications
Antigen | Humanin |
---|---|
Concentration | 1 mg/mL |
Dilution | Western Blot 1:500 |
Applications | Western Blot |
Classification | Polyclonal |
Description
Humanin Polyclonal specifically detects Humanin in Human samples. It is validated for Western Blot.Specifications
Humanin | |
Western Blot 1:500 | |
Polyclonal | |
Rabbit | |
Human | |
HN, HUNIN | |
MTRNR2L1 | |
IgG | |
Affinity Purified | |
This is specific for Humanin. |
1 mg/mL | |
Western Blot | |
Unconjugated | |
RUO | |
PBS with 0.02% Sodium Azide | |
100462977 | |
Residues 9-24 (LLLLTSEIDLPVKRRA) of the human Humanin protein. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title