Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Humanin Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10056877
Description
Humanin Polyclonal specifically detects Humanin in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| Humanin | |
| Polyclonal | |
| Western Blot 1:500-1:2000, ELISA 1:100-1:1000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence reported by customer review, Immunohistochemistry-Paraffin 1:100 | |
| Q8IVG9 | |
| MTRNR2L1 | |
| The full length peptide sequence from the human MT-RNR2 protein (accesion number Q8IVG9). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Antisera |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Whole antisera with 50% Glycerol with 0.025% Sodium Azide | |
| HN, HUNIN | |
| Rabbit | |
| Unpurified | |
| RUO | |
| 100462977 | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction