Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Humanin Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.50 - $427.50
Specifications
| Antigen | Humanin |
|---|---|
| Dilution | Western Blot 1:500-1:2000, ELISA 1:100-1:1000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence reported by customer review, Immunohistochemistry-Paraffin 1:100 |
| Applications | Western Blot, ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB10056877T
![]() |
Novus Biologicals
NB10056877SS |
0.025 mL |
Each for $206.50
|
|
|||||
NB10056877
![]() |
Novus Biologicals
NB10056877 |
0.1 mL |
Each for $427.50
|
|
|||||
Description
Humanin Polyclonal specifically detects Humanin in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| Humanin | |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q8IVG9 | |
| 100462977 | |
| The full length peptide sequence from the human MT-RNR2 protein (accesion number Q8IVG9). | |
| Primary | |
| Store at -20C. Avoid freeze-thaw cycles. |
| Western Blot 1:500-1:2000, ELISA 1:100-1:1000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence reported by customer review, Immunohistochemistry-Paraffin 1:100 | |
| Polyclonal | |
| Antisera | |
| RUO | |
| Whole antisera with 50% Glycerol with 0.025% Sodium Azide | |
| HN, HUNIN | |
| MTRNR2L1 | |
| IgG | |
| Unpurified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title