Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HUNK Antibody, Novus Biologicals™
SDP

Catalog No. NBP214110 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
25ul
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP214110 0.1 mL
NB438079 25ul
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP214110 Supplier Novus Biologicals Supplier No. NBP214110
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HUNK Polyclonal specifically detects HUNK in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen HUNK
Applications Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:200 - 1:500
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Alias B19, EC 2.7.11, EC 2.7.11.1, hormonally up-regulated neu tumor-associated kinase, hormonally upregulated neu tumor-associated kinase, hormonally up-regulated Neu-associated kinase, hormonally upregulated Neu-associated kinase, MAKV, serine/threonine protein kinase MAK-V, Serine/threonine-protein kinase MAK-V
Gene Symbols HUNK
Host Species Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to the amino acids: SSFPDKDSFGCRNIFRKTSDSNCVASSSMEFIPVPPPRTPRIVKKPEPHQPGPGSTGIPHKEDPLMLDMVRSFESVDR
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, GPCR, Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 30811
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.