Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Huntingtin Antibody, HRP, Novus Biologicals™
SDP

Catalog No. NB120806 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB120806 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB120806 Supplier Novus Biologicals Supplier No. NBP144266H
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Huntingtin Polyclonal antibody specifically detects Huntingtin in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry

Specifications

Antigen Huntingtin
Applications Western Blot, ELISA, Immunohistochemistry
Classification Polyclonal
Conjugate HRP
Dilution Western Blot, ELISA, Immunohistochemistry
Formulation PBS
Gene Alias HD protein, huntingtin, huntingtin (Huntington disease), IT15HDHuntington disease protein
Host Species Rabbit
Immunogen Huntingtin Antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to the C-terminus of human Huntingtin disease protein. (Uniprot: P42858)
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Autophagy, Cell Biology, Cellular Markers, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 3064
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.