Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HuR/ELAVL1 Rabbit anti-Human, Mouse, Rat, Clone: JM96-33, Novus Biologicals™
SDP

Catalog No. NBP275532 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP275532 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP275532 Supplier Novus Biologicals Supplier No. NBP275532

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Rabbit Monoclonal Antibody

HuR/ELAVL1 Monoclonal specifically detects HuR/ELAVL1 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunoprecipitation, Immunohistochemistry-Paraffin.

Specifications

Antigen HuR/ELAVL1
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone JM96-33
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin
Formulation TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide
Gene Alias ELAV (embryonic lethal, abnormal vision, Drosophila)-like 1 (Hu antigen R), ELAV-like protein 1, embryonic lethal, abnormal vision, drosophila, homolog-like 1, Hu antigen R, Hua, Hu-antigen R, HuR, HURELAV1, MelG
Gene Symbols ELAVL1
Host Species Rabbit
Immunogen Synthetic peptide within Human HuR/ELAVL1 aa 50-99/ 326. (SwissProt: Q15717 Human; SwissProt: P70372 Mouse; SwissProt: B5DF91 Rat)
Molecular Weight of Antigen 36 kDa
Purification Method Protein A purified
Quantity 100 μL
Research Discipline Vision
Primary or Secondary Primary
Gene ID (Entrez) 1994.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.