Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HUS1B Antibody, Novus Biologicals™
SDP

Catalog No. NBP177749 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP177749 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP177749 Supplier Novus Biologicals Supplier No. NBP177749
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HUS1B Polyclonal specifically detects HUS1B in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation.

Specifications

Antigen HUS1B
Applications Western Blot, ELISA, Immunoprecipitation
Classification Polyclonal
Concentration 1.75 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1:500-1:5000, ELISA 1:20000-1:100000, Immunoprecipitation 1:100
Formulation 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide
Gene Accession No. Q8NHY5
Gene Alias checkpoint protein HUS1B, hHUS1B, HUS1 (S. pombe) checkpoint homolog b, HUS1 checkpoint homolog b (S. pombe), MGC126746, MGC126748, RP11-532F6.1
Gene Symbols HUS1B
Host Species Rabbit
Immunogen This affinity purified HUS1B Antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to an internal region near amino acids 100-125 of human HUS1B protein. (Uniprot: Q8NHY5)
Molecular Weight of Antigen 30.98 kDa
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 135458
Test Specificity This antibody is directed against human Hus1B protein. A BLAST analysis indicates 100% identity of the immunizing sequence with Hus1B protein.
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.