Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HUWE1 Antibody, Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NB100652AFX Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB100652AFX 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB100652AFX Supplier Novus Biologicals Supplier No. NB100652AF647
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HUWE1 Polyclonal specifically detects HUWE1 in Human, Mouse samples. It is validated for Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin).

Specifications

Antigen HUWE1
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 647
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias EC 6.3.2, EC 6.3.2.6, HECT, UBA and WWE domain containing 1, HECTH9, HSPC272, KIAA0312MULE, Mcl-1 ubiquitin ligase E3, Mule, UREB1UBA and WWE domain-containing protein 1, URE-binding protein 1
Gene Symbols HUWE1
Host Species Rabbit
Immunogen The immunogen this antibody was made to, maps to a region between residues 2250 and 2300 of human Upstream Regulatory Element Binding Protein 1 using the numbering given in entry NP_113584.3 (GeneID 10075).
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 10075
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.