Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HUWE1 Antibody (AX81), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP229784
Description
HUWE1 Monoclonal specifically detects HUWE1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
HUWE1 | |
Monoclonal | |
Unconjugated | |
Ascites with 0.1% Sodium Azide | |
EC 6.3.2, EC 6.3.2.6, HECT, UBA and WWE domain containing 1, HECTH9, HSPC272, KIAA0312MULE, Mcl-1 ubiquitin ligase E3, Mule, UREB1UBA and WWE domain-containing protein 1, URE-binding protein 1 | |
Mouse | |
480 kDa | |
0.1 mL | |
Stem Cell Markers | |
10075 | |
Store at 4C. Do not freeze. |
Immunocytochemistry, Immunofluorescence | |
AX81 | |
Immunocytochemistry/Immunofluorescence 1:10-1:2000 | |
Q7Z6Z7 | |
HUWE1 | |
Synthetic peptide cooresponding to amino acids 2530-2610 of human HectH9 (Adhikary, S. et al, 2005). | |
Unpurified | |
RUO | |
Primary | |
Human | |
Ascites |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction