Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HUWE1 Antibody (AX81), Novus Biologicals™

Mouse Monoclonal Antibody
Specifications
Antigen | HUWE1 |
---|---|
Clone | AX81 |
Dilution | Immunocytochemistry/Immunofluorescence 1:10-1:2000 |
Applications | Immunocytochemistry, Immunofluorescence |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP229784
![]() |
Novus Biologicals
NBP229784 |
0.1 mL | N/A | N/A | N/A | ||||
Description
HUWE1 Monoclonal specifically detects HUWE1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
HUWE1 | |
Immunocytochemistry/Immunofluorescence 1:10-1:2000 | |
Monoclonal | |
Ascites | |
RUO | |
Human | |
Q7Z6Z7 | |
10075 | |
Synthetic peptide cooresponding to amino acids 2530-2610 of human HectH9 (Adhikary, S. et al, 2005). | |
Unpurified | |
480 kDa |
AX81 | |
Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
Mouse | |
Stem Cell Markers | |
Ascites with 0.1% Sodium Azide | |
EC 6.3.2, EC 6.3.2.6, HECT, UBA and WWE domain containing 1, HECTH9, HSPC272, KIAA0312MULE, Mcl-1 ubiquitin ligase E3, Mule, UREB1UBA and WWE domain-containing protein 1, URE-binding protein 1 | |
HUWE1 | |
Primary | |
Store at 4C. Do not freeze. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title