Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HUWE1 Antibody (BLR129H), Novus Biologicals™
SDP

Catalog No. NB188052 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB188052 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB188052 Supplier Novus Biologicals Supplier No. NBP314712
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

HUWE1 Monoclonal antibody specifically detects HUWE1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)

Specifications

Antigen HUWE1
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone BLR129H
Conjugate Unconjugated
Dilution Western Blot 1:1000, Immunohistochemistry 1:100 - 1:500, Immunocytochemistry/Immunofluorescence 1:100 - 1:500, Immunoprecipitation 20 ul/1 mg lysate, Immunohistochemistry-Paraffin 1:100 - 1:500
Formulation Borate Buffered Saline (BBS) pH 8.2 with 0.1% BSA and 0.09% Sodium Azide
Gene Alias arf-bp1, EC 6.3.2, EC 6.3.2.6, HECT, UBA and WWE domain containing 1, HECTH9, HSPC272, KIAA0312MULE, Mcl-1 ubiquitin ligase E3, Mule, UREB1UBA and WWE domain-containing protein 1, URE-binding protein 1
Host Species Rabbit
Immunogen between 2250 and 2300
Quantity 100 μg
Regulatory Status RUO
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 10075
Target Species Human, Mouse
Content And Storage Store at 2 - 8°C/ 1 year from date of receipt
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.