Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HVEM/TNFRSF14 Antibody (2024B), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB348923 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB348923 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB348923 Supplier Novus Biologicals Supplier No. FAB2516MFV610
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

HVEM/TNFRSF14 Monoclonal specifically detects HVEM/TNFRSF14 in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen HVEM/TNFRSF14
Applications Flow Cytometry
Classification Monoclonal
Clone 2024B
Conjugate mFluor Violet 610 SE
Dilution Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias CD270 antigen, CD40-like protein, Herpes virus entry mediator A, Herpesvirus entry mediator A, HveA, HVEMTR2HVEAATAR, LIGHTR, tumor necrosis factor receptor superfamily member 14, tumor necrosis factor receptor superfamily, member 14 (herpesvirus entrymediator), Tumor necrosis factor receptor-like 2, tumor necrosis factor receptor-like gene2
Host Species Rabbit
Immunogen Chinese hamster ovary cell line CHO-derived recombinant mouse HVEM/TNFRSF14, Gln39-Val207, Accession # NP_849262
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, B Cell Development and Differentiation Markers, Cancer, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 8764
Target Species Mouse
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.