Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HVEM/TNFRSF14 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP176690 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP176690 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP176690 Supplier Novus Biologicals Supplier No. NBP176690
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HVEM/TNFRSF14 Polyclonal specifically detects HVEM/TNFRSF14 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunofluorescence.

Specifications

Antigen HVEM/TNFRSF14
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1-2 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 1 μg/mL, Immunocytochemistry/Immunofluorescence 10 μg/mL, Immunohistochemistry-Paraffin 1 μg/mL
Formulation PBS with 0.02% Sodium Azide
Gene Accession No. Q92956
Gene Alias CD270 antigen, CD40-like protein, Herpes virus entry mediator A, Herpesvirus entry mediator A, HveA, HVEMTR2HVEAATAR, LIGHTR, tumor necrosis factor receptor superfamily member 14, tumor necrosis factor receptor superfamily, member 14 (herpesvirus entrymediator), Tumor necrosis factor receptor-like 2, tumor necrosis factor receptor-like gene2
Gene Symbols TNFRSF14
Host Species Rabbit
Immunogen Antibody was raised against a 15 amino acid synthetic peptide from near the amino terminus of human TNFRSF14. The immunogen is located within amino acids 20 - 70 of TNFRSF14. Amino Acid Squence: PSCREDEYPVGSECC
Molecular Weight of Antigen 23 kDa
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 8764
Test Specificity Multiple isoforms of TNFRSF14 are known to exist.
Target Species Human, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.