Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HVEM/TNFRSF14 Antibody, FITC, Novus Biologicals™
SDP

Catalog No. NB198060 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB198060 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB198060 Supplier Novus Biologicals Supplier No. NBP176690F
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HVEM/TNFRSF14 Polyclonal antibody specifically detects HVEM/TNFRSF14 in Human, Mouse samples. It is validated for ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen HVEM/TNFRSF14
Applications ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate FITC
Formulation PBS
Gene Alias CD270 antigen, CD40-like protein, Herpes virus entry mediator A, Herpesvirus entry mediator A, HveA, HVEMTR2HVEAATAR, LIGHTR, tumor necrosis factor receptor superfamily member 14, tumor necrosis factor receptor superfamily, member 14 (herpesvirus entrymediator), Tumor necrosis factor receptor-like 2, tumor necrosis factor receptor-like gene2
Host Species Rabbit
Immunogen Antibody was raised against a 15 amino acid synthetic peptide from near the amino terminus of human TNFRSF14. The immunogen is located within amino acids 20 - 70 of TNFRSF14. Amino Acid Squence: PSCREDEYPVGSECC
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, B Cell Development and Differentiation Markers, Cancer, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 8764
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.